missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylhydroquinone, 97%

Product Code. 10582281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10582281 5 g Glass bottle 5g
10219553 25 g Glass bottle 25g
11914681 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10582281 Supplier Thermo Scientific Acros Supplier No. 305110050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-43-5 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-43-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00009997
InChI Key BXJGUBZTZWCMEX-UHFFFAOYSA-N
Synonym 2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone
PubChem CID 69100
IUPAC Name 2,3-dimethylbenzene-1,4-diol
SMILES CC1=C(C)C(O)=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 138.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 3-Dimethylhydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.