missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzonitrile, 97%, Thermo Scientific™

Product Code. 11444663
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444663 5 g 5g
11454663 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444663 Supplier Thermo Scientific Alfa Aesar Supplier No. A11589.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5724-56-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00019744
InChI Key ZEDPQIJYJCPIRM-UHFFFAOYSA-N
Synonym benzonitrile, 2,3-dimethyl, 3-cyano-o-xylene, unii-6s215f38ok, 2,3-dimethylbenzonitril german, 2,3-dimethylbenzenecarbonitrile, 2,3-dimethylbenzonitril, dimethylbenzonitrile, pubchem8125, benzonitrile, dimethyl, medetomidine impurity 11
PubChem CID 79802
IUPAC Name 2,3-dimethylbenzonitrile
SMILES CC1=CC=CC(C#N)=C1C

Spécification

Melting Point 23°C to 25°C
Quantity 5 g
Odor Almond-like
Refractive Index 1.532
Beilstein 1932620
Boiling Point 64°C to 66°C (0.8 mmHg)
Formula Weight 131.18
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylbenzonitrile
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.