missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzoic acid, 98%

Product Code. 11418563
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418563 100 g 100g
11498553 5 g 5g
11408563 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418563 Supplier Thermo Scientific Alfa Aesar Supplier No. A10383.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 603-79-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-79-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002479
InChI Key RIZUCYSQUWMQLX-UHFFFAOYSA-N
Synonym hemellitic acid, benzoic acid, 2,3-dimethyl, 2,3-dimethylbenzoicacid, 2,3-dimethyl benzoic acid, 2,3-dimethyl-benzoic acid, unii-7irp8ca267, 2,3-dimethylbenzenecarboxylic acid, vic-o-xylylic acid, methyl m-toluic acid, pubchem2549
PubChem CID 11782
ChEBI CHEBI:64823
IUPAC Name 2,3-dimethylbenzoic acid
SMILES CC1=CC=CC(=C1C)C(=O)O

Specifications

Melting Point 143°C to 147°C
Quantity 100 g
Odor Odorless
Beilstein 971590
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.