missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylaniline-d3, TRC

Product Code. 30384304
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384304 50 mg
30387717 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30384304 Supplier Toronto Research Chemicals Supplier No. TRCD46219250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57414-78-5
Molecular Formula C8 2H3 H8 N
Molecular Weight (g/mol) 124.2
Synonym 2,3-Dimethylaniline D3
IUPAC Name 3-methyl-2-(trideuteriomethyl)aniline
SMILES [2H]C([2H])([2H])c1c(C)cccc1N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H11N/c1-6-4-3-5-8(9)7(6)2/h3-5H,9H2,1-2H3/i2D3
Formula Weight 124.108
Chemical Name or Material 2,3-Dimethylaniline-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.