missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylaniline, 99%, Thermo Scientific™

Product Code. 10520741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10520741 5mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10520741 Supplier Thermo Scientific Acros Supplier No. 115930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 87-59-2
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Molecular Formula C8H11N
Linear Formula (CH3)2C6H3NH2
MDL Number MFCD00007732
Beilstein 12, 1101
Merck Index 15, 10282
Synonym 2,3-xylidine, 3-amino-o-xylene, benzenamine, 2,3-dimethyl, 2,3-dimethylbenzenamine, 2,3-dimethylphenylamine, 2,3-xylylamine, ortho-xylidine, vic-o-xylidine, unii-zd450abi9x, ccris 4739
Solubility Information Solubility in water: slightly soluble 30g/L (20°C). Other solubilities: soluble in many organic solvents,,very soluble in ethanol and diethyl ether,ccl4,,dmso
InChI Key VVAKEQGKZNKUSU-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)N)C
IUPAC Name 2,3-dimethylaniline
Molecular Weight (g/mol) 121.18
PubChem CID 6893
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2, 3-Dimethylaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.