missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Dimethylamino)propyl]-1,2-dihydro-1-(phenylmethyl)-3H-indazol-3-one Hydrochloride, TRC

Product Code. 30372827
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372827 25 mg -
30373634 100 mg -
30387474 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372827 Supplier Toronto Research Chemicals Supplier No. TRCD48213025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85284-06-6
Molecular Formula C19H23N3O . HCl
Molecular Weight (g/mol) 309.4
Synonym 1-Benzyl-2-[3-(dimethylamino)propyl]-1,2-dihydro-3H-indazol-3-one Hydrochloride, 3H-Indazol-3-one, 2-[3-(dimethylamino)propyl]-1,2-dihydro-1-(phenylmethyl)-, monohydrochloride (9CI)
SMILES CN(C)CCCN1N(Cc2ccccc2)c3ccccc3C1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H23N3O/c1-20(2)13-8-14-21-19(23)17-11-6-7-12-18(17)22(21)15-16-9-4-3-5-10-16/h3-7,9-12H,8,13-15H2,1-2H3
Formula Weight 345.1608
Chemical Name or Material 2-[3-(Dimethylamino)propyl]-1,2-dihydro-1-(phenylmethyl)-3H-indazol-3-one Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.