missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, ≥97%, Thermo Scientific™

Product Code. 10071874
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071874 250 mg Amber Glass Bottle 250mg
10071824 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071874 Supplier Thermo Scientific Maybridge Supplier No. CC59646CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 910037-06-8
Molecular Formula C13H22N2O
Molecular Weight (g/mol) 222.332
MDL Number MFCD09064998
InChI Key PSLNWMDOKBMVNV-UHFFFAOYSA-N
Synonym 2-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-2-methylamino methyl phenoxy propyl amine, 2-3-dimethylamino propoxy phenyl methyl methyl amine, n,n-dimethyl-3-2-methylamino methyl phenoxy propan-1-amine, benzenemethanamine,2-3-dimethylamino propoxy-n-methyl
PubChem CID 24229613
IUPAC Name N,N-dimethyl-3-[2-(methylaminomethyl)phenoxy]propan-1-amine
SMILES CNCC1=CC=CC=C1OCCCN(C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.33
Percent Purity 97%
Chemical Name or Material 2-[3-(Dimethylamino)propoxy]-N-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.