missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethyl-quinoxaline-6-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
IUPAC Name | 2,3-dimethylquinoxaline-6-carboxylate |
Formula Weight | 202.21 |
Percent Purity | 97% |
Chemical Name or Material | 2, 3-Dimethyl-quinoxaline-6-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'