missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-N-phenylbenzenamine, TRC

Product Code. 30388222
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388222 2.5 g 2.5g
30377010 1 g 1g
30381080 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388222 Supplier Toronto Research Chemicals Supplier No. TRCD4874052.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4869-11-8
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
Synonym N-Phenyl-2,3-dimethylaniline, Benzenamine, 2,3-dimethyl-N-phenyl-, 2,3-Xylidine, N-phenyl- (7CI,8CI)
SMILES Cc1cccc(Nc2ccccc2)c1C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H15N/c1-11-7-6-10-14(12(11)2)15-13-8-4-3-5-9-13/h3-10,15H,1-2H3
Formula Weight 197.1204
Chemical Name or Material 2,3-Dimethyl-N-phenylbenzenamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.