missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-6-nitro-2H-indazole, TRC

Product Code. 30387465
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387465 5 g
30381692 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387465 Supplier Toronto Research Chemicals Supplier No. TRCD4710555G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444731-73-1
Molecular Formula C9 H9 N3 O2
Molecular Weight (g/mol) 191.19
Synonym 2H-Indazole, 2,3-dimethyl-6-nitro-, 2,3-Dimethyl-6-nitro-2H-indazole
IUPAC Name 2,3-dimethyl-6-nitroindazole
SMILES Cc1c2ccc(cc2nn1C)[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H9N3O2/c1-6-8-4-3-7(12(13)14)5-9(8)10-11(6)2/h3-5H,1-2H3
Formula Weight 191.0695
Chemical Name or Material 2,3-Dimethyl-6-nitro-2H-indazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.