missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-5H,6H,7H,8H-imidazo[1,2-a]pyrazine (~85%), TRC

Product Code. 30387959
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387959 100 mg
30384105 10 mg
30381347 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387959 Supplier Toronto Research Chemicals Supplier No. TRCD475430100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1250515-54-8
Molecular Formula C8H13N3
Molecular Weight (g/mol) 151.21
Synonym 2,3-Dimethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine, 5,6,7,8-Tetrahydro-2,3-dimethylimidazo[1,2-a]pyrazine
IUPAC Name 2,3-dimethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine
SMILES Cc1nc2CNCCn2c1C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H13N3/c1-6-7(2)11-4-3-9-5-8(11)10-6/h9H,3-5H2,1-2H3
Formula Weight 151.1109
Chemical Name or Material 2,3-Dimethyl-5H,6H,7H,8H-imidazo[1,2-a]pyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.