missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2-butene, 98%

Product Code. 10574454 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
Product Code. Quantity Packaging unitSize
10574454 500 mL Glass bottle 500mL
10061690 25 mL Glass bottle 25mL
10734281 100 mL Glass bottle 100mL
3 options
This item is not returnable. View return policy

Product Code. 10574454

Brand: Thermo Scientific Acros 156595000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 563-79-1 | C6H12 | 84.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-79-1
Molecular Formula C6H12
Molecular Weight (g/mol) 84.15
MDL Number MFCD00008897
InChI Key WGLLSSPDPJPLOR-UHFFFAOYSA-N
Synonym 2,3-dimethyl-2-butene, tetramethylethylene, 2-butene, 2,3-dimethyl, 2,3-dimethylbutene-2, 1,1,2,2-tetramethylethylene, tetramethylethene, 2,3-dimethyl-but-2-ene, ch3 2c=c ch3 2, 2-butene,3-dimethyl, acmc-1akps
PubChem CID 11250
IUPAC Name 2,3-dimethylbut-2-ene
SMILES CC(=C(C)C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -75.0°C
Color Colorless
Density 0.7000g/mL
Boiling Point 73.0°C
Flash Point −16°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=C(CH3)2
Refractive Index 1.4110 to 1.4130
Quantity 500 mL
Beilstein 01, 218
Specific Gravity 0.7
Formula Weight 84.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethyl-2-butene
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethyl-2-butene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.