missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-2,3-diphenylbutane, 95%

Product Code. 10796042 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10796042

Brand: Thermo Scientific Acros 349981000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1889-67-4 | C18H22 | 238.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1889-67-4
Molecular Formula C18H22
Molecular Weight (g/mol) 238.37
MDL Number MFCD00053713
InChI Key HGTUJZTUQFXBIH-UHFFFAOYSA-N
Synonym 2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene
PubChem CID 74681
IUPAC Name (2,3-dimethyl-3-phenylbutan-2-yl)benzene
SMILES CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 90°C to 110°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Linear Formula C6H5C(CH3)2C(CH3)2C6H5
Quantity 100 g
Beilstein 05,396
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in toluene
Formula Weight 238.37
Percent Purity 95%
Physical Form Crystalline Powder, Flakes, or Chunks
Chemical Name or Material 2, 3-Dimethyl-2, 3-diphenylbutane, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.