missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1-butene, 99%

Product Code. 11317545
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317545 25 g 25g
11307545 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317545 Supplier Thermo Scientific Alfa Aesar Supplier No. L05675.14
  • ONLINE EXCLUSIVE PRICE
    £120.36
    £103.65 / 25g
    Save £16.71
    13% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 563-78-0 | C6H12 | 84.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-78-0
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00008923
InChI Key OWWIWYDDISJUMY-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1-butene, 1-butene, 2,3-dimethyl, 2,3-dimethylbutene, 2,3-dimethylbutene-1, 2,3-dimethyl butene, acmc-1aqwm, butene, 2,3-dimethyl, ch3 2chc ch3 =ch2, 1-butene, 2,3-dimethyl-8ci 9ci
PubChem CID 11249
IUPAC Name 2,3-dimethylbut-1-ene
SMILES CC(C)C(=C)C

Specifications

Melting Point -157°C
Density 0.679
Boiling Point 56°C to 57°C
Flash Point −18°C (−°F)
Refractive Index 1.391
Quantity 25 g
UN Number UN3295
Beilstein 505982
Formula Weight 84.16
Percent Purity 99%
Chemical Name or Material 2,3-Dimethyl-1-butene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.