missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1-butene, 99%

Product Code. 11307545
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11307545

Brand: Thermo Scientific Alfa Aesar L05675.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 563-78-0 | C6H12 | 84.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-78-0
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00008923
InChI Key OWWIWYDDISJUMY-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1-butene, 1-butene, 2,3-dimethyl, 2,3-dimethylbutene, 2,3-dimethylbutene-1, 2,3-dimethyl butene, acmc-1aqwm, butene, 2,3-dimethyl, ch3 2chc ch3 =ch2, 1-butene, 2,3-dimethyl-8ci 9ci
PubChem CID 11249
IUPAC Name 2,3-dimethylbut-1-ene
SMILES CC(C)C(=C)C

Specifications

Melting Point -157°C
Boiling Point 56°C to 57°C
Flash Point −18°C (−°F)
Quantity 5 g
UN Number UN3295
Beilstein 505982
Formula Weight 84.16
Percent Purity 99%
Chemical Name or Material 2,3-Dimethyl-1-butene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.