missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1,3-butadiene, 98%, stab. with 100ppm BHT

Product Code. 11326846
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326846 50 g 50g
11316846 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326846 Supplier Thermo Scientific Alfa Aesar Supplier No. L04207.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 513-81-5 | C6H10 | 82.15 g/mol

2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.

Solubility
Miscible with chloroform. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-81-5
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00008595
InChI Key SDJHPPZKZZWAKF-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene
PubChem CID 10566
IUPAC Name 2,3-dimethylbuta-1,3-diene
SMILES CC(=C)C(C)=C

Specifications

Melting Point -76°C
Density 0.726
Boiling Point 68°C to 69°C
Flash Point −1°C (30°F)
Refractive Index 1.438
Quantity 50 g
UN Number UN3295
Beilstein 605285
Merck Index 14,3238
Solubility Information Miscible with chloroform. Immiscible with water.
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 2,3-Dimethyl-1,3-butadiene, stabilized with 100ppm BHT
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.