missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxytoluene, 98+%

Product Code. 11484703
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484703 5 g 5g
11494703 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484703 Supplier Thermo Scientific Alfa Aesar Supplier No. A11617.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4463-33-6 | C9H12O2 | 152.193 g/mol

2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.

Solubility
Soluble in Dichloromethane. Not miscible in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4463-33-6
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00008379
InChI Key WMXFNCKPYCAIQW-UHFFFAOYSA-N
Synonym 2,3-dimethoxytoluene, 3-methylveratrole, benzene, 1,2-dimethoxy-3-methyl, dimethoxytoluene, 1,2-dimethoxy-3-methyl-benzene, acmc-1ae12, 1-methyl-2,3-dimethoxybenzene, 3-methylcatechol, dimethyl ether, #
PubChem CID 78215
IUPAC Name 1,2-dimethoxy-3-methylbenzene
SMILES CC1=C(C(=CC=C1)OC)OC

Specifications

Density 1.024
Boiling Point 202°C to 203°C
Flash Point 85°C (185°F)
Odor Aromatic
Refractive Index 1.514
Quantity 5 g
Beilstein 1940962
Solubility Information Soluble in Dichloromethane. Not miscible in water.
Formula Weight 152.19
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxytoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.