missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxyphenol, 98%

Product Code. 15431957
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431957 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15431957 Supplier Thermo Scientific Alfa Aesar Supplier No. 044060.04
 Restricted Item
Estimated Shipment: 01-12-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 5150-42-5 | C8H10O3 | 154.165 g/mol

2,3-Dimethoxyphenol is used to study the nitrosative deamination of DNA bases induced by reactive nitrogen species (RNS) while studying the cause of mutagenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 5150-42-5
Density 1.182 g/mL
Boiling Point 233°C to 234°C
Flash Point 108°C (226°F)
Molecular Formula C8H10O3
Refractive Index 1.54
MDL Number MFCD00008366
Quantity 2 g
Synonym dimethoxyphenol, phenol, dimethoxy, unii-ow147i6c84, 1-hydroxy-2,3-dimethoxybenzene, pyrogallol 1,2-dimethyl ether, phenol, 2,3-dimethoxy, phenol,3-dimethoxy, 2,3-di-methoxyphenol, 2,3-dim ethoxyphenol, 2,3-dimethoxy phenol
Solubility Information Soluble in Chloroform.
InChI Key QSZCGGBDNYTQHH-UHFFFAOYSA-N
SMILES COC1=CC=CC(=C1OC)O
IUPAC Name 2,3-dimethoxyphenol
Molecular Weight (g/mol) 154.165
PubChem CID 78828
Formula Weight 154.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethoxyphenol
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.