missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxynaphthalene, 97%, Thermo Scientific Chemicals

Product Code. 15411818
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Refrigerated (2-4°C). Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10103-06-7
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD05664308
InChI Key XYRPWXOJBNGTMX-UHFFFAOYSA-N
PubChem CID 604708
IUPAC Name 2,3-dimethoxynaphthalene
SMILES COC1=CC2=CC=CC=C2C=C1OC

Specifications

Melting Point 116°C to 121°C
Color White to Yellow
Quantity 5 g
Synonym naphthalene, 2,3-dimethoxy, acmc-20aft8, naphthalene,2,3-dimethoxy, 2,3-dimethoxynaphthalene, 2,3-dihydroxynaphthalene, dimethyl ether
Formula Weight 188.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2,3-Dimethoxynaphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'