missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzylamine, 99%, Thermo Scientific™

Product Code. 10555100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10555100 5 g Glass bottle 5g
10565100 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10555100 Supplier Thermo Scientific Acros Supplier No. 433390050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4393-09-3
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00052392
InChI Key LVMPWFJVYMXSNY-UHFFFAOYSA-N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 140.0°C to 142.0°C (15.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.11
Synonym 2,3-dimethoxybenzylamine, 2,3-dimethoxyphenyl methanamine, 1-2,3-dimethoxyphenyl methanamine, benzenemethanamine, 2,3-dimethoxy, 2,3-dimethoxy-benzylamine, 2,3-dimethoxy benzylamine, pubchem7399, 2,3-dimethoxybenzyl amine, acmc-2097fc
SMILES COC1=CC=CC(=C1OC)CN
IUPAC Name (2,3-dimethoxyphenyl)methanamine
PubChem CID 78106
Formula Weight 167.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.