missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimercapto-1-propanol, 97.0-100.5%

Product Code. 10685722 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10685722 5 mL Glass bottle 5mL
10722184 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10685722 Supplier Thermo Scientific Acros Supplier No. 115300050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-52-9 | C3H8OS2 | 124.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-52-9
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.22
MDL Number MFCD00004864
InChI Key WQABCVAJNWAXTE-UHFFFAOYSA-N
Synonym dimercaprol, 2,3-dimercapto-1-propanol, 2,3-dimercaptopropanol, dithioglycerine, dicaptol, sulfactin, dimercaptopropanol, british anti-lewisite, 2,3-dithiopropanol, antoxol
PubChem CID 3080
ChEBI CHEBI:64198
IUPAC Name 2,3-bis(sulfanyl)propan-1-ol
SMILES C(C(CS)S)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2430g/mL
Boiling Point 120.0°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.0-100.5%
Packaging Glass bottle
Linear Formula HSCH2CH(SH)CH2OH
Refractive Index 1.5700 to 1.5740
Quantity 5 mL
Merck Index 15,3231
Specific Gravity 1.243
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol and vegetable oils
Formula Weight 124.22
Percent Purity 97.0-100.5%
Physical Form Liquid
Chemical Name or Material 2,3-Dimercapto-1-propanol, 97.0-100.5%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.