missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%

Product Code. 15442776
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15442776

Brand: Thermo Scientific Alfa Aesar H56578.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207233-91-8 | C3H7O3S3 | 187.27 g/mol

Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.

Solubility
Soluble in Water: 100 mg/ml

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207233-91-8
Molecular Formula C3H7O3S3
Molecular Weight (g/mol) 187.27
MDL Number MFCD00149543
InChI Key JLVSRWOIZZXQAD-GSVOUGTGSA-M
Synonym sodium 2,3-dimercaptopropanesulfonate monohydrate, unithiol, dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropane-1-sulfonate hydrate, 2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropanesulfonate hydrate, dmps, sodium 2,3-disulfanylpropane-1-sulfonate hydrate, unitiol monohydrate
PubChem CID 23676755
SMILES [O-]S(=O)(=O)C[C@H](S)CS

Specifications

Melting Point ∽229°C (decomposition)
Quantity 250 mg
Beilstein 3734863
Merck Index 14,3210
Solubility Information Soluble in Water: 100mg/ml
IUPAC Name (2R)-2,3-disulfanylpropane-1-sulfonate
Formula Weight 228.30 (210.28 Anhydrous)
Percent Purity 98%
Chemical Name or Material 2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.