missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 99.95% (metals basis)

Product Code. 11404110
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404110 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404110 Supplier Thermo Scientific Alfa Aesar Supplier No. A11307.09
  • ONLINE EXCLUSIVE PRICE
    £22.54
    £18.65 / 10g
    Save £3.89
    17% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-44-4 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2,3-Dihydroxynaphthalene is used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 92-44-4
Melting Point 162°C to 165°C
Flash Point 175°C
Molecular Formula C10H8O2
MDL Number MFCD00004073
Quantity 10 g
Beilstein 742375
Synonym 2,3-dihydroxynaphthalene, 2,3-naphthalenediol, 2,3-dihydroxynapthalene, 2,3-dihydroxy naphthalene, 2,3-dihydroxynaphthlene
Solubility Information Slightly soluble aq. soln.
InChI Key JRNGUTKWMSBIBF-UHFFFAOYSA-N
SMILES OC1=C(O)C=C2C=CC=CC2=C1
IUPAC Name naphthalene-2,3-diol
Molecular Weight (g/mol) 160.17
PubChem CID 7091
ChEBI CHEBI:38135
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material 2,3-Dihydroxynaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.