missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxybenzoic acid, 98%

Product Code. 11469412
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469412 50 g 50g
11459412 10 g 10g
11479412 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469412 Supplier Thermo Scientific Alfa Aesar Supplier No. A10144.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 303-38-8 | C7H6O4 | 154.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Dihydroxybenzoic acids are used as intermediates for pharmaceuticals and other organic synthesis. Used as matrix for ionization of peptides, proteins and carbohydrates. In industrial field used for resins, polyesters, plasticizers, dyestuffs, preservatives, and rubber chemicals. It is also used as is a selected phenol as MMP (Matrix Metalloproteinase) inhibitor.

Solubility
Soluble in water, methanol and dimethyl sulfoxide.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-38-8
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002446
InChI Key GLDQAMYCGOIJDV-UHFFFAOYSA-N
Synonym pyrocatechuic acid, o-pyrocatechuic acid, 2-pyrocatechuic acid, 3-hydroxysalicylic acid, dobk, dhba, 2,3-dihydroxybenzoicacid, benzoic acid, 2,3-dihydroxy, catecholcarboxylic acid, 2,3-dihydroxy-benzoic acid
PubChem CID 19
ChEBI CHEBI:18026
IUPAC Name 2,3-dihydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)O)O)C(=O)O

Specifications

Melting Point 204°C to 208°C
Density 1.542
Quantity 50 g
Beilstein 2209117
Solubility Information Soluble in water,methanol and dimethyl sulfoxide.
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material 2,3-Dihydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.