missing translation for 'onlineSavingsMsg'
Learn More

2,3-dihydroxy-3-methylbutanoic acid, TRC

Product Code. 30388227
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388227 10 mg
30369279 100 mg
30384365 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388227 Supplier Toronto Research Chemicals Supplier No. TRCD45605510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1756-18-9
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
IUPAC Name 2,3-dihydroxy-3-methylbutanoic acid
SMILES CC(C)(O)C(O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C5H10O4/c1-5(2,9)3(6)4(7)8/h3,6,9H,1-2H3,(H,7,8)
Formula Weight 134.0579
Chemical Name or Material 2,3-dihydroxy-3-methylbutanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.