missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10235624
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10235624 1 g Amber Glass Bottle 1g
10641475 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10235624 Supplier Thermo Scientific Maybridge Supplier No. CC47801DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 260063-21-6
Molecular Formula C7H6O4S
Molecular Weight (g/mol) 186.181
MDL Number MFCD07801102
InChI Key DCCRIIFBJZOPAV-UHFFFAOYSA-N
Synonym 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylicacid, 2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carboxylic acid, 3,4-ethylenedioxythiophene carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylic acid, 2,3-dihydro
PubChem CID 6483880
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
SMILES C1COC2=C(SC=C2O1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 186.19
Percent Purity 97%
Chemical Name or Material 2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.