missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydropyrido[2,3-d][1,3]oxazol-2-one, 97%, Thermo Scientific™

Catalog Number 10345012
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
60832-72-6
C6H4N2O2
136.11
MFCD00204215,MFCD09065038
OVLXOTUWFLHWQT-UHFFFAOYSA-N
oxazolo 4,5-b pyridin-2 3h-one, 3h-oxazolo 4,5-b pyridin-2-one, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 1,3 oxazolo 4,5-b pyridin-2 3h-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, pubchem11061, 2-oxooxazolo 4,5-b pyridine
2799900
CHEBI:38580
3H-[1,3]oxazolo[4,5-b]pyridin-2-one
O=C1NC2=NC=CC=C2O1
This item is not returnable. View return policy
60832-72-6
C6H4N2O2
136.11
MFCD00204215,MFCD09065038
OVLXOTUWFLHWQT-UHFFFAOYSA-N
oxazolo 4,5-b pyridin-2 3h-one, 3h-oxazolo 4,5-b pyridin-2-one, 2,3-dihydropyrido 2,3-d 1,3 oxazol-2-one, 1,3 oxazolo 4,5-b pyridin-2-ol, 1,3 oxazolo 4,5-b pyridin-2 3h-one, 3h-1,3 oxazolo 4,5-b pyridin-2-one, 2h,3h-1,3 oxazolo 4,5-b pyridin-2-one, pubchem11061, 2-oxooxazolo 4,5-b pyridine
2799900
CHEBI:38580
3H-[1,3]oxazolo[4,5-b]pyridin-2-one
O=C1NC2=NC=CC=C2O1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 136.11
Percent Purity 97%
Chemical Name or Material [1, 3]Oxazolo[4, 5-b]pyridin-2-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'