missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-7-ylamine, 97%, Thermo Scientific™

Product Code. 10071664
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071664 1 g Amber Glass Bottle 1g
10404665 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071664 Supplier Thermo Scientific Maybridge Supplier No. CC00914DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13414-56-7
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD11109313
InChI Key UHHZGSLXPQGPJL-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-7-amine, 2,3-dihydrobenzo b furan-7-ylamine, 2,3-dihydro-benzofuran-7-ylamine, 7-benzofuranamine, 2,3-dihydro, 7-amino-2,3-dihydrobenzofuran, 2,3-dihydro-7-benzofuranamine
PubChem CID 12805950
IUPAC Name 2,3-dihydro-1-benzofuran-7-amine
SMILES NC1=CC=CC2=C1OCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-7-ylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.