missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-5-benzofuranacetic Acid-d2, TRC

Product Code. 30380477
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30380477 Supplier Toronto Research Chemicals Supplier No. TRCD44848750MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dihydro-5-benzofuranacetic Acid-d2
CAS 1216878-16-8
Molecular Formula C10H8D2O3
Quantity 50 mg
Synonym (2,3-Dihydrobenzofuran-5-yl)acetic Acid-d2, 2,3-Dihydrobenzofuran-5-acetic Acid-d2, 2-(2,3-Dihydrobenzofuran-5-yl)acetic Acid-d2
InChI Formula InChI=1S/C10H10O3/c11-10(12)6-7-1-2-9-8(5-7)3-4-13-9/h1-2,5H,3-4,6H2,(H,11,12)/i6D2
SMILES [2H]C([2H])(C(=O)O)c1ccc2OCCc2c1
IUPAC Name 2,2-dideuterio-2-(2,3-dihydro-1-benzofuran-5-yl)acetic acid
Molecular Weight (g/mol) 180.2
Formula Weight 180.0755
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.