missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylic Acid tert-Butyl Ester, TRC

Product Code. 30387926
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387926 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387926 Supplier Toronto Research Chemicals Supplier No. TRCD44981025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylic Acid tert-Butyl Ester
CAS 837392-67-3
Molecular Formula C19H28BNO4
Quantity 25 mg
Synonym tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline-1-carboxylate 2,3-Dihydro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C19H28BNO4/c1-17(2,3)23-16(22)21-11-10-13-12-14(8-9-15(13)21)20-24-18(4,5)19(6,7)25-20/h8-9,12H,10-11H2,1-7H3
SMILES CC(C)(C)OC(=O)N1CCc2cc(ccc12)B3OC(C)(C)C(C)(C)O3
Molecular Weight (g/mol) 345.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.