missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-2-methyl-1H-indol-1-amine Hydrochloride, TRC

Product Code. 30382187
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382187 50 g
30373394 1 g
30371841 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382187 Supplier Toronto Research Chemicals Supplier No. TRCD44964550G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 102789-79-7
Molecular Formula C9 H12 N2 . Cl H
Molecular Weight (g/mol) 184.67
Synonym 1H-Indol-1-amine, 2,3-dihydro-2-methyl-, hydrochloride (9CI), 1-Amino-2-methylindoline Hydrochloride
IUPAC Name 2-methyl-2,3-dihydroindol-1-amine;hydrochloride
SMILES Cl.CC1Cc2ccccc2N1N

Specifikationer

Quantity 50 g
InChI Formula InChI=1S/C9H12N2.ClH/c1-7-6-8-4-2-3-5-9(8)11(7)10;/h2-5,7H,6,10H2,1H3;1H
Formula Weight 184.0767
Chemical Name or Material 2-Methylindolin-1-amine hydrochloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.