missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1H-indol-2-ylmethanol, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 149.19 |
Percent Purity | 97% |
Chemical Name or Material | 2, 3-Dihydro-1H-indol-2-ylmethanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'