missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-7-ylmethylamine, 97%, Thermo Scientific™

Product Code. 10061664
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061664 1 g Amber Glass Bottle 1g
10346464 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10061664 Supplier Thermo Scientific Maybridge Supplier No. CC00913DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 361393-65-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD11109312
InChI Key LNQLTDVDXCZARO-UHFFFAOYSA-N
Synonym 2,3-dihydro-1-benzofuran-7-ylmethylamine, 1-2,3-dihydro-1-benzofuran-7-yl methanamine, 7-benzofuranmethanamine, 2,3-dihydro, 2,3-dihydrobenzofuran-7-yl methanamine, 2,3-dihydrobenzofuran-7-yl methyl amine, 2,3-dihydro-1-benzo b furan-7-ylmethylamine, 2,3-dihydrobenzo 2,3-b furan-7-ylmethylamine
PubChem CID 23512832
IUPAC Name 2,3-dihydro-1-benzofuran-7-ylmethanamine
SMILES C1COC2=C1C=CC=C2CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 149.19
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-7-ylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.