missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-yl Isocyanate, TRC

Product Code. 30394569
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30394569 10 mg 10mg
30391367 100 mg -
30390848 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394569 Supplier Toronto Research Chemicals Supplier No. TRCI77941810MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215162-92-8
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
IUPAC Name 5-isocyanato-2,3-dihydro-1-benzofuran
SMILES O=C=Nc1ccc2OCCc2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H7NO2/c11-6-10-8-1-2-9-7(5-8)3-4-12-9/h1-2,5H,3-4H2
Formula Weight 161.0477
Chemical Name or Material 2,3-Dihydro-1-benzofuran-5-yl Isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.