missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10407232
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10407232 1 g 1g
11320557 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10407232 Supplier Thermo Scientific Maybridge Supplier No. CC00803DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 115010-11-2
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.65
MDL Number MFCD02677703
InChI Key RVWYPBARHGPULM-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-sulfonyl chloride, 2,3-dihydro-1-benzofuran-5-sulfonoylchloride, 2,3-dihydro-1-benzofuran-5-sulfonoyl chloride, 2,3-dihydrobenzofuran-5-sulfonylchloride, 2,3-dihydro-benzofuran-5-sulfonyl chloride, 2,3-dihydrobenzo b furan-5-sulfonyl chloride, pubchem14874, 5-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzofuran-5-sulfonylchloride, chloro-2,3-dihydrobenzo b furan-5-ylsulfone
PubChem CID 2776154
IUPAC Name 2,3-dihydro-1-benzofuran-5-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC2=C(OCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 218.66
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.