missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2696912
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1914-60-9
C9H8O3
164.16
MFCD03086168
WEVFUSSJCGAVOH-UHFFFAOYNA-N
2,3-dihydrobenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 2,3-dihydro, 2,3-dihydro-benzofuran-2-carboxylic acid, 2,3-dihydrobenzo b furan-2-carboxylic acid, 2-benzofurancarboxylicacid, 2,3-dihydro, 2,3-dihydro-2-benzofurancarboxylic acid, dihydrobenzofuran-2-carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylic, 2,3-dihydrobenzofuran carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylicacid
2776555
2,3-dihydro-1-benzofuran-2-carboxylic acid
OC(=O)C1CC2=CC=CC=C2O1
This item is not returnable. View return policy
1914-60-9
C9H8O3
164.16
MFCD03086168
WEVFUSSJCGAVOH-UHFFFAOYNA-N
2,3-dihydrobenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 2,3-dihydro, 2,3-dihydro-benzofuran-2-carboxylic acid, 2,3-dihydrobenzo b furan-2-carboxylic acid, 2-benzofurancarboxylicacid, 2,3-dihydro, 2,3-dihydro-2-benzofurancarboxylic acid, dihydrobenzofuran-2-carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylic, 2,3-dihydrobenzofuran carboxylic acid, 2,3-dihydrobenzofuran-2-carboxylicacid
2776555
2,3-dihydro-1-benzofuran-2-carboxylic acid
OC(=O)C1CC2=CC=CC=C2O1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'