missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-2-carbonyl chloride, 95%, Thermo Scientific™

Product Code. 10743813
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10743813 1 g Amber Glass Bottle 1g
10733813 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10743813 Supplier Thermo Scientific Maybridge Supplier No. CC23902DA
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 27347-32-6
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD03086169
InChI Key QZVPHVMVWZKULP-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-2-carbonyl chloride, 2-benzofurancarbonylchloride, 2,3-dihydro, 2,3-dihydrobenzo b furan-2-carbonyl chloride, 2,3-dihydro-benzofuran-2-carbonyl chloride, 2-benzofurancarbonyl chloride, 2,3-dihydro-7ci,8ci,9ci
PubChem CID 2795427
IUPAC Name 2,3-dihydro-1-benzofuran-2-carbonyl chloride
SMILES C1C(OC2=CC=CC=C21)C(=O)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 182.61
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.