missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-5-carbonitrile, 97%, Thermo Scientific™

Product Code. 10473993
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10473993

Brand: Thermo Scientific Maybridge CC01416DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 148703-14-4
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD01116986
InChI Key WNNQCWLSQDNACP-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b 1,4 dioxine-5-carbonitrile, 1,4-benzodioxin-5-carbonitrile,2,3-dihydro, acmc-1c2ut, 2h,3h-benzo e 1,4-dioxane-5-carbonitrile
PubChem CID 2795043
IUPAC Name 2,3-dihydro-1,4-benzodioxine-5-carbonitrile
SMILES C1COC2=C(C=CC=C2O1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-5-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.