missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxine-2-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250mg
1g
5g
Especificaciones
Assay Percent Range | 97% |
Quantity | 5g |
Formula Weight | 180.16 |
Percent Purity | 97% |
Chemical Name or Material | 2, 3-Dihydro-1, 4-benzodioxine-2-carboxylic acid |