missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxin-5-ylmethylamine, 97%, Thermo Scientific™

Product Code. 10051234
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051234 5 g Amber Glass Bottle 5g
10640903 250 mg Amber Glass Bottle 250mg
10170582 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051234 Supplier Thermo Scientific Maybridge Supplier No. CC01413DE
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 261633-71-0
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
InChI Key OKRXRKUGZVMPKX-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine
PubChem CID 2795040
IUPAC Name 2,3-dihydro-1,4-benzodioxin-5-ylmethanamine
SMILES C1COC2=C(C=CC=C2O1)CN

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 201.65
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-5-ylmethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.