missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11986481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11986481 5 g 5g
11976481 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986481 Supplier Thermo Scientific Acros Supplier No. 446770050
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 851386-31-7 | Molecular Formula: C6H3F2NO2 | Molecular Weight: 159.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851386-31-7
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
InChI Key FFGHMEKFPCGYEG-UHFFFAOYSA-N
Synonym 2,3-difluoroisonicotinic acid, 2,3-difluoro-4-pyridinecarboxylic acid, 2,3-difluoro-4-carboxypyridine, 4-pyridinecarboxylic acid, 2,3-difluoro, 2,3-fifluoroisonicotinic acid, 4-carboxy-2,3-difluoropyridine, 2,3-difluoroisonicotinicacid, pubchem18431, ksc447o0d, 5,6-difluoropyridine-4-carboxylic acid
PubChem CID 11240637
IUPAC Name 2,3-difluoropyridine-4-carboxylic acid
SMILES C1=CN=C(C(=C1C(=O)O)F)F

Specifications

Melting Point 165°C to 167°C
Assay Percent Range 97%
Quantity 5 g
Formula Weight 159.09
Percent Purity 97%
Chemical Name or Material 2, 3-Difluoropyridine-4-carboxylic acid
compliance-icons
Product Identifier
  • 2,3-Difluoropyridine-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.