missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenylboronic acid, 97+%, Thermo Scientific™

Product Code. 10660165 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber glass bottle
Glass bottle
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10660165 1 g Amber glass bottle 1g
10252852 1 g Glass bottle 1g
10212902 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10660165 Supplier Thermo Scientific Maybridge Supplier No. AC35912DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121219-16-7
MDL Number MFCD01863170
InChI Key SZYXKFKWFYUOGZ-UHFFFAOYSA-N
Synonym 2,3-difluorobenzeneboronic acid, 2,3-difluorophenyl boronic acid, 2,3-difluorophenyl boranediol, 2,3-difluorophenylboronicacid, 2,3-difluoro phenylboric acid, 2,3-fluorobenzene boronic acid, 2,3-difluoro phenylboronic acid, 2,3-difluorophenyloboric acid, boronic acid, 2,3-difluorophenyl
PubChem CID 2734333
IUPAC Name (2,3-difluorophenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)F)F)(O)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Amber glass bottle
Molecular Formula C6H5BF2O2
Quantity 1 g
Molecular Weight (g/mol) 157.911
Formula Weight 157.91
Percent Purity ≥97.0%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Difluorophenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.