missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenylacetic acid, 97%, ACROS Organics™

Brand:  Acros Organics 435470010

Code : 29

Additional Details : CAS Number : 145689-41-4 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 10381280

  • £21.90 / 1g


Estimated Shipment
14-06-2018
Add to basket


Specifications

Chemical Identifiers

2, 3-Difluorophenylacetic acid
96.0
97%
C8H6F2O2
MFCD00040968
145689-41-4
100.0
97%
172.13

Specifications

2, 3-Difluorophenylacetic acid
117.0° to 119.0°C
Authentic
96.0
Crystalline Powder
97%
172.13
White to off-white
1g
145689-41-4
100.0
97%
C8H6F2O2
MFCD00040968
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,3-Difluorophenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS