missing translation for 'onlineSavingsMsg'
Learn More

2,3-(Difluoromethylenedioxy)benzoic acid, Dr. Ehrenstorfer

Product Code. 30311084
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311084 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311084 Supplier Dr. Ehrenstorfer Supplier No. DREC13705030
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fludioxonil-despyrrol carboxylic acid
CAS 126120-85-2
Analyte or Component Names Fludioxonil-despyrrol carboxylic acid
Shipping Condition Room Temperature
Molecular Formula C8 H4 F2 O4
Certifications/Compliance ISO 17034
Synonym 1,3-Benzodioxole-4-carboxylic acid, 2,2-difluoro-, 2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid, 2,2-Difluorobenzo[d][1,3]dioxole-4-carboxylic acid, 2,3-(Difluoromethylenedioxy)benzoic acid, Fludioxonil-despyrrol carboxylic acid
InChI Formula InChI=1S/C8H4F2O4/c9-8(10)13-5-3-1-2-4(7(11)12)6(5)14-8/h1-3H,(H,11,12)
Solution Type Neat
SMILES OC(=O)c1cccc2OC(F)(F)Oc12
IUPAC Name 2,2-difluoro-1,3-benzodioxole-4-carboxylic acid
Industry Type Environmental Testing
Molecular Weight (g/mol) 202.11
Quantity 50 mg
Formula Weight 202.0078 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.