missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 11479666
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11479666

Brand: Thermo Scientific Alfa Aesar B20214.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113211-94-2
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD00042488
InChI Key FTBSGSZZESQDBM-UHFFFAOYSA-N
PubChem CID 517984
IUPAC Name 1-(bromomethyl)-2,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)F)CBr

Specifications

Density 1.628
Flash Point 90°C (194°F)
Refractive Index 1.5298
Quantity 5 g
UN Number UN1760
Beilstein 7089244
Synonym 2,3-difluorobenzyl bromide, 1-bromomethyl-2,3-difluorobenzene, alpha-bromo-2,3-difluorotoluene, benzene, 1-bromomethyl-2,3-difluoro, 2,3-difluorobenzylbromide, a-bromo-2,3-difluorotoluene, 1-bromomethyl-2,3-difluoro-benzene, 2,3-difluorophenylbromide, 2, 3-difluorobenzyl bromide, pubchem4909
Formula Weight 207.02
Percent Purity 97%
Chemical Name or Material 2,3-Difluorobenzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.