missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorobenzyl alcohol, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
3.9mL
Specifications
Specifications
| CAS | 75853-18-8 |
| CAS Min % | 92.0 |
| CAS Max % | 100.0 |
| Color | Colorless to Brown |
| Density | 1.2800g/mL |
| Flash Point | 93°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Molecular Formula | C7H6F2O |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction