missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorobenzoyl chloride, 98%, Thermo Scientific™
Click to view available options
1g; Glass bottle
Specifications
CAS | 18355-73-2 |
Density | 1.4230g/mL |
Boiling Point | 85.0°C to 87.0°C (14.0mmHg) |
Flash Point | 91°C |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C7H3ClF2O |
MDL Number | MFCD00042499 |
Quantity | 1g |
Specific Gravity | 1.423 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'