missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzonitrile, 99%

Product Code. 11445736
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11445736

Brand: Thermo Scientific Alfa Aesar B20811.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21524-39-0 | C7H3F2N | 139.105 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21524-39-0
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.105
MDL Number MFCD00009976
InChI Key GKPHNZYMLJPYJJ-UHFFFAOYSA-N
Synonym benzonitrile, 2,3-difluoro, 2,3-difluorobenzenecarbonitrile, ncr bf cf, difluorobenzonitrile, pubchem1564, acmc-209flg, 2,3-difluorobenzonitril, 2,3-difluoro-benzonitrile, 2.3-difluoro benzonitrile, ksc493a8b
PubChem CID 88935
IUPAC Name 2,3-difluorobenzonitrile
SMILES C1=CC(=C(C(=C1)F)F)C#N

Specifications

Density 1.254
Boiling Point 187°C to 188°C
Flash Point 75°C (167°F)
Odor Almond-like
Refractive Index 1.4882
Quantity 5 g
UN Number UN3276
Beilstein 2085621
Formula Weight 139.11
Percent Purity 99%
Chemical Name or Material 2,3-Difluorobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.