missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzaldehyde, 98%, Thermo Scientific™

Product Code. 10793151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10793151 1 g Glass bottle 1g
10177513 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10793151 Supplier Thermo Scientific Acros Supplier No. 303510010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 2646-91-5
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010292
InChI Key WDBAXYQUOZDFOJ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3-difluoro, 2,3-difluoro benzaldehyde, 2,3-difluoro-benzaldehyde, difluoro-benzaldehyde, pubchem1436, 2,3 difluorobenzaldehyde, acmc-1cr6r, 2,3-difluorobenzaldehyde?, 2,3-difluorobenzaldehyde, ksc493k8r
PubChem CID 137664
IUPAC Name 2,3-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)F)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.3000g/mL
Boiling Point 64°C to 65°C (17.0 mmHg)
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CHO
Refractive Index 1.4980 to 1.5
Quantity 1 g
Specific Gravity 1.3
Formula Weight 142.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3-Difluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.