missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Difluoroacetophenone, 97%

Product Code. 11462808
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462808 5 g 5g
11452808 1 g 1g
11472808 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462808 Supplier Thermo Scientific Alfa Aesar Supplier No. A16786.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18355-80-1 | C8H6F2O | 156.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18355-80-1
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00061274
InChI Key PQUXFUBNSYCQAL-UHFFFAOYSA-N
Synonym 2',3'-difluoroacetophenone, 1-2,3-difluorophenyl ethanone, 2,3-difluoroacetophenone, 1-2,3-difluorophenyl ethan-1-one, ethanone, 1-2,3-difluorophenyl, 1-acetyl-2,3-difluorobenzene, pubchem4211, acmc-209x0g, ksc184c5n, 2',3'-difluoro acetophenone
PubChem CID 519550
SMILES CC(=O)C1=CC=CC(F)=C1F

Specifications

Boiling Point 84°C to 85°C (20 mmHg)
Odor Characteristic
Refractive Index 1.489
Quantity 5 g
Beilstein 2963572
IUPAC Name 1-(2,3-difluorophenyl)ethanone
Formula Weight 156.13
Percent Purity 97%
Chemical Name or Material 2',3'-Difluoroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.